Anti-IFI6 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313901EA 564 EUR
ORIGTA313901
Anti-IFI6 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:1000; ELISA: 1:10000

Type: Primary
Antigen: IFI6 (interferon, alpha-inducible protein 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR