Anti-B4GALNT1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313910EA 564 EUR
ORIGTA313910
Anti-B4GALNT1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:5000

Type: Primary
Antigen: B4GALNT1 (beta-1,4-N-acetyl-galactosaminyl transferase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR