Anti-GNAZ Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313934EA 564 EUR
ORIGTA313934
Anti-GNAZ Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:5000

Type: Primary
Antigen: GNAZ (guanine nucleotide binding protein (G protein), alpha z polypeptide)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR