Anti-GRM8 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA313976
ORIGTA313976EA 564 EUR
ORIGTA313976
Anti-GRM8 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:100-1:1000; Immunochemistry: 1:50-1:100; ELISA: 1:10000

Type: Primary
Antigen: GRM8 (glutamate receptor, metabotropic 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR