Anti-MSH6 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313980EA 552 EUR
ORIGTA313980
Anti-MSH6 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot:1.500-1.3000; Immunochemistry: 1:50-1:100; ELISA: 1:5000

Type: Primary
Antigen: MSH6 (mutS homolog 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR