Anti-GTF2H1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313983EA 552 EUR
ORIGTA313983
Anti-GTF2H1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: GTF2H1 (general transcription factor IIH, polypeptide 1, 62kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR