Anti-HNRNPD Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314021EA 564 EUR
ORIGTA314021
Anti-HNRNPD Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: HNRNPD (heterogeneous nuclear ribonucleoprotein D (AU-rich element RNA binding protein 1, 37kDa))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR