Anti-HTR1E Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314038EA 543 EUR
ORIGTA314038
Anti-HTR1E Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:1000

Type: Primary
Antigen: HTR1E (5-hydroxytryptamine (serotonin) receptor 1E, G protein-coupled)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR