Anti-ITPKB Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314103EA 552 EUR
ORIGTA314103
Anti-ITPKB Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: ITPKB (inositol-trisphosphate 3-kinase B)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR