Anti-JUNB Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314117EA 532 EUR
ORIGTA314117
Anti-JUNB Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:20000; Immunoprecipitation: Various Dilution

Type: Primary
Antigen: JUNB (jun B proto-oncogene)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR