Anti-KCNC2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314124EA 543 EUR
ORIGTA314124
Anti-KCNC2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:10000

Type: Primary
Antigen: KCNC2 (potassium voltage-gated channel, Shaw-related subfamily, member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR