Anti-LAMA5 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314146EA 532 EUR
ORIGTA314146
Anti-LAMA5 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: LAMA5 (laminin, alpha 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR