Anti-LGALS4 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314152EA 564 EUR
ORIGTA314152
Anti-LGALS4 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunochemistry: 1:50-1:100 elisa: 1:20000

Type: Primary
Antigen: LGALS4 (lectin, galactoside-binding, soluble, 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR