Anti-MYBL1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314222EA 564 EUR
ORIGTA314222
Anti-MYBL1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: MYBL1 (v-myb avian myeloblastosis viral oncogene homolog-like 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR