Anti-MYL6 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314227EA 552 EUR
ORIGTA314227
Anti-MYL6 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:1000

Type: Primary
Antigen: MYL6 (myosin, light chain 6, alkali, smooth muscle and non-muscle)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR