Anti-MYO5A Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314229EA 564 EUR
ORIGTA314229
Anti-MYO5A Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: MYO5A (myosin VA (heavy chain 12, myoxin))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR