Anti-NDUFV3 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314242EA 552 EUR
ORIGTA314242
Anti-NDUFV3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: NDUFV3 (NADH dehydrogenase (ubiquinone) flavoprotein 3, 10kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR