Anti-NKTR Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314263EA 532 EUR
ORIGTA314263
Anti-NKTR Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:5000

Type: Primary
Antigen: NKTR (natural killer cell triggering receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR