Anti-NOS2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314265EA 552 EUR
ORIGTA314265
Anti-NOS2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: NOS2 (nitric oxide synthase 2, inducible)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR