Anti-GPR143 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314295EA 564 EUR
ORIGTA314295
Anti-GPR143 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:10000

Type: Primary
Antigen: GPR143 (G protein-coupled receptor 143)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR