Anti-P2RY4 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314314EA 564 EUR
ORIGTA314314
Anti-P2RY4 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: P2RY4 (pyrimidinergic receptor P2Y, G-protein coupled, 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR