Anti-PKIA Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314383EA 543 EUR
ORIGTA314383
Anti-PKIA Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: PKIA (protein kinase (cAMP-dependent, catalytic) inhibitor alpha)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR