Anti-RASGRF1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314421EA 552 EUR
ORIGTA314421
Anti-RASGRF1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:1000

Type: Primary
Antigen: RASGRF1 (Ras protein-specific guanine nucleotide-releasing factor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR