Anti-RFX2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314441EA 532 EUR
ORIGTA314441
Anti-RFX2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:10000

Type: Primary
Antigen: RFX2 (regulatory factor X, 2 (influences HLA class II expression))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR