Anti-SCN9A Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314513EA 552 EUR
ORIGTA314513
Anti-SCN9A Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: SCN9A (sodium channel, voltage-gated, type IX, alpha subunit)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR