Anti-SLC2A3 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314539EA 543 EUR
ORIGTA314539
Anti-SLC2A3 Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:1000

Type: Primary
Antigen: SLC2A3 (solute carrier family 2 (facilitated glucose transporter), member 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR