Anti-TBX1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314594EA 552 EUR
ORIGTA314594
Anti-TBX1 Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: TBX1 (T-box 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR