Anti-TCEB3 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314597EA 564 EUR
ORIGTA314597
Anti-TCEB3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:1000

Type: Primary
Antigen: TCEB3 (transcription elongation factor B (SIII), polypeptide 3 (110kDa, elongin A))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR