Anti-ZNF148 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314693EA 564 EUR
ORIGTA314693
Anti-ZNF148 Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:1000

Type: Primary
Antigen: ZNF148 (zinc finger protein 148)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR