Anti-FZD5 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314704EA 564 EUR
ORIGTA314704
Anti-FZD5 Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions: Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:5000

Type: Primary
Antigen: FZD5 (frizzled class receptor 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR