Anti-DDX3Y Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314783EA 532 EUR
ORIGTA314783
Anti-DDX3Y Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: DDX3Y (DEAD (Asp-Glu-Ala-Asp) box helicase 3, Y-linked)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR