Anti-CDKL1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314812EA 552 EUR
ORIGTA314812
Anti-CDKL1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: CDKL1 (cyclin-dependent kinase-like 1 (CDC2-related kinase))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR