Anti-RAB7L1 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA314835
ORIGTA314835EA 564 EUR
ORIGTA314835
Anti-RAB7L1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: RAB7L1 (RAB7, member RAS oncogene family-like 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR