Anti-SLC16A3 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314860EA 564 EUR
ORIGTA314860
Anti-SLC16A3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: SLC16A3 (solute carrier family 16 (monocarboxylate transporter), member 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR