Anti-GPR50 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314884EA 552 EUR
ORIGTA314884
Anti-GPR50 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:10000

Type: Primary
Antigen: GPR50 (G protein-coupled receptor 50)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR