Anti-TP53I11 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA314930EA 532 EUR
ORIGTA314930
Anti-TP53I11 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: TP53I11 (tumor protein p53 inducible protein 11)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR