Anti-CA14 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA315264
ORIGTA315264EA 543 EUR
ORIGTA315264
Anti-CA14 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:5000

Type: Primary
Antigen: CA14 (carbonic anhydrase XIV)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR