Anti-IL1RAPL2 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA315322
ORIGTA315322EA 564 EUR
ORIGTA315322
Anti-IL1RAPL2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: IL1RAPL2 (interleukin 1 receptor accessory protein-like 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR