Anti-OR10H3 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA315330EA 564 EUR
ORIGTA315330
Anti-OR10H3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: OR10H3 (olfactory receptor, family 10, subfamily H, member 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR