Anti-OR7C2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA315339EA 564 EUR
ORIGTA315339
Anti-OR7C2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: OR7C2 (olfactory receptor, family 7, subfamily C, member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR