Anti-LAMP3 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA315363EA 564 EUR
ORIGTA315363
Anti-LAMP3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: LAMP3 (lysosomal-associated membrane protein 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR