Anti-HTR5BP Rabbit Polyclonal Antibody

Lieferant: OriGene
TA315779
ORIGTA315779EA 543 EUR
ORIGTA315779
Anti-HTR5BP Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:40000

Type: Primary
Antigen: HTR5BP (5-hydroxytryptamine (serotonin) receptor 5B, pseudogene)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR