Anti-COL23A1 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA315979
ORIGTA315979EA 543 EUR
ORIGTA315979
Anti-COL23A1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:10000

Type: Primary
Antigen: COL23A1 (collagen, type XXIII, alpha 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR