Anti-LDLRAD3 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA316138EA 543 EUR
ORIGTA316138
Anti-LDLRAD3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:10000

Type: Primary
Antigen: LDLRAD3 (low density lipoprotein receptor class A domain containing 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR