Anti-C1QL2 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA316193
ORIGTA316193EA 564 EUR
ORIGTA316193
Anti-C1QL2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: C1QL2 (complement component 1, q subcomponent-like 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR