Anti-ZNF596 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA316199
ORIGTA316199EA 543 EUR
ORIGTA316199
Anti-ZNF596 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:1000

Type: Primary
Antigen: ZNF596 (zinc finger protein 596)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR