Anti-TAS2R20 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA316281
ORIGTA316281EA 564 EUR
ORIGTA316281
Anti-TAS2R20 Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: TAS2R20 (taste receptor, type 2, member 20)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR