Anti-OR10G4 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA316435
ORIGTA316435EA 543 EUR
ORIGTA316435
Anti-OR10G4 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: OR10G4 (olfactory receptor, family 10, subfamily G, member 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR