Anti-DRD2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA316680EA 680 EUR
ORIGTA316680
Anti-DRD2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 3-4 µg/ml

Type: Primary
Antigen: DRD2 (dopamine receptor D2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR