Anti-GRPR Rabbit Polyclonal Antibody

Lieferant: OriGene
TA316873
ORIGTA316873EA 680 EUR
ORIGTA316873
Anti-GRPR Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 5 µg/ml

Type: Primary
Antigen: GRPR (gastrin-releasing peptide receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR